Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Diethylamino-1-propanol | 622-93-5 | MFCD00002947 | 98.0% | 131.22 | 1 ML
SDP

Catalog No. 5000366446
Encompass_Preferred

3-Diethylamino-1-propanol is a tertiary amine compound recognized for its anticonvulsant activity. This chemical is supplied as a colorless to light yellow liquid, suitable for laboratory research applications.

  • Anticonvulsant activity
  • Liquid appearance
  • Colorless to light yellow
  • Molecular weight of 131.22
  • Soluble in DMSO (100 mg/mL)
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Melting/freezing point: -12.63 °C
  • Boiling point: 81-83°C at 15 mmHg
  • Flash point: 65.6 °C
  • Relative density: 0.884 g/cm³
  • For research use only

Catalog No. 50-003-66446 Supplier Medchemexpress LLC Supplier No. HYW01038310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Diethylamino-1-propanol is a tertiary amine compound recognized for its anticonvulsant activity. This chemical is supplied as a colorless to light yellow liquid, suitable for laboratory research applications.

  • Anticonvulsant activity
  • Liquid appearance
  • Colorless to light yellow
  • Molecular weight of 131.22
  • Soluble in DMSO (100 mg/mL)
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Melting/freezing point: -12.63 °C
  • Boiling point: 81-83°C at 15 mmHg
  • Flash point: 65.6 °C
  • Relative density: 0.884 g/cm³
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.