Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Diethylamino-1-propanol 95.0+%, TCI America™
SDP

Catalog No. D0472500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0472500ML 500 mL
D047225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0472500ML Supplier TCI America Supplier No. D0472500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 622-93-5
Molecular Formula C7H17NO
Molecular Weight (g/mol) 131.22
MDL Number MFCD00002947
InChI Key WKCYFSZDBICRKL-UHFFFAOYSA-N
Synonym 3-diethylamino-1-propanol, 3-diethylamino propan-1-ol, 1-propanol, 3-diethylamino, 3-diethylamino propanol, 3-diethylamino propyl alcohol, n,n-diethyl-3-hydroxypropylamine, diethylpropanolamine, unii-u57yow5553, gamma-diaethylaminopropanol german
PubChem CID 12159
IUPAC Name 3-(diethylamino)propan-1-ol
SMILES CCN(CC)CCCO

Specifications

Color Colorless
Boiling Point 189°C
Quantity 500 mL
Formula Weight 131.22
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Diethylamino-1-propanol
RTECSNumber UB1480000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.