Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [3-(Diethylamino)Propyl]Trimethoxysilane

Catalog No. 4486645ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-866-45ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 44-866-45ML Supplier Sigma Aldrich Supplier No. 4486645ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[3-(Diethylamino)propyl]trimethoxysilane was covalently bonded to mesoporous titania–silica mixed oxides to act as epoxidation catalysts.{55} [3-(Diethylamino)propyl]trimethoxysilane, immobilized on mesoporous silicas (SBA-15) may be used as adsorbents to capture CO2. {56}

Specifications

CAS 79069-13-9
Melting Point 59°C to 62°C
Molecular Formula C8H17NO3
Refractive Index n20/D 1.423
Linear Formula CH3CH[NHCO2C(CH3)3]CH2OH
MDL Number MFCD00043121
Quantity 5 mL
Synonym N,N-Diethyl-3-(trimethoxysilyl)propylamine
Molecular Weight (g/mol) 235.4 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.