Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Diethylamino)propylamine

Catalog No. 502891515 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-1515 2 L
54-997-5500ML 500 mL
54-997-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-289-1515 Supplier Sigma Aldrich Supplier No. 5499752L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-(diethylamino)propylamine (3-(diethylamino)-1-propylamine, deapa) is a diamine.3-(diethylamino)propylamine may be used in the preparation of:
  • Biocompatible polyampholyte-coated (poly(acrylic acid)-co-3-(diethylamino)-propylamine magnetite nanoparticles (pamnps)
  • Hyaluronic acid grafted with 3-diethylaminopropyl (deap) group (ha-g-deap) nanoparticles for use as a drug delivery vehicle
  • Deapa-modified poly(vinyl alcohol) (pva) polymers that can be employed in gene silencing.it may also be used to introduce cationic groups during the surface modification of microfibrillated cellulose (mfc).product of arkema inc
TRUSTED_SUSTAINABILITY

Specifications

CAS 104-78-9
Density 0.826 g/mL (at 25°C (literature))
Boiling Point 159°C (lit.)
Molecular Formula C7H18N2
Refractive Index n20/D 1.4416 (literature)
Linear Formula (C2H5)2 N(CH2)3 NH2
MDL Number MFCD00008218
Quantity 2 L
Synonym N,N-Diethyl-1,3-diaminopropane; N,N-Diethyl-1,3-propanediamine; DEAPA
Molecular Weight (g/mol) 130.23
Percent Purity ≥99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.