Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Diethylaminopropylamine, 99+%

Catalog No. AC114090025
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114090025 2.5 L Glass bottle
AC114092500 250 mL Glass bottle
AC114090010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC114090025 Supplier Thermo Scientific Chemicals Supplier No. 114090025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-78-9 | C7H18N2 | 130.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-78-9
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00008218
InChI Key QOHMWDJIBGVPIF-UHFFFAOYSA-N
Synonym 3-diethylaminopropylamine, 3-diethylamino propylamine, n,n-diethyl-1,3-propanediamine, 3-aminopropyl diethylamine, n1,n1-diethylpropane-1,3-diamine, 1,3-propanediamine, n,n-diethyl, diethylaminotrimethylenamine, n,n-diethyl-1,3-diaminopropane, 1-diethylamino propylamine-3, n-3-diethylaminopropyl amine
PubChem CID 61011
IUPAC Name N',N'-diethylpropane-1,3-diamine
SMILES CCN(CC)CCCN

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -60°C
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 168°C to 171°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4406 to 1.4426
Linear Formula (C2H5)2N(CH2)3NH2
Quantity 2.5 L
Specific Gravity 0.82
Solubility Information Solubility in water: miscible with water
Viscosity 1.3 mPa.s (20°C)
Formula Weight 130.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Diethylaminopropylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-236-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.