Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Difluoromethoxy)phenol

Catalog No. CDS005791 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005791 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005791 Supplier Sigma Aldrich Supplier No. CDS005791100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-ethylacrolein can be used as a promoter to synthesize 2-alkylchroman-4-ones via rhodium-catalyzed annulation reaction of 1,3-dienes with salicylaldehydes or 2-hydroxybenzyl alcohols.it can also be used as a reactant:
  • In the total synthesis of (+/-)-tabersonine, an indole alkaloid of the aspidosperma family
  • To prepare 2-ethylbicyclo[2.2.1]hept-5-ene-2-carboxaldehyde via enantioselective diels-alder reaction with cyclopentadiene in the presence of chiral lewis acid catalyst
  • To synthesize diethyl 5-ethyl-2,3-pyridinedicarboxylate by reacting with 2-chloro-3-oxo-succinic acid diethyl ester using ammonium acetate as a catalyst

Specifications

Molecular Formula C7H6F2O2
Linear Formula C7H6F2O2
MDL Number MFCD00236235
Quantity 5 g
Molecular Weight (g/mol) 160.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.