Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Dimethoxy)-5-(tributylstannyl)pyridine

Catalog No. SYX000721G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX000721G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX000721G Supplier Sigma Aldrich Supplier No. SYX000721G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,6-dimethylcyclohexanone, mixture of isomers is a clear, light yellow liquid
  • The adsorption of traces of 2,6-dimethylcyclohexanone, a volatile organic compound, from liquid toluene was studied
  • Cis- and trans-isomers of 2,6-dimethylcyclohexanonereacts with hydroxylamine hydrochloride and potassium hydroxide in methanol solution gives cis- and trans-dimethylcyclohexanone oxime .2,6-dimethylcyclohexanone was used in tetraoxane synthesis and in evaluation of their anti-malarial activity

Specifications

CAS 1264193-67-0
Molecular Formula C20H37NO2Sn
Linear Formula C20H37NO2Sn
MDL Number MFCD10699129
Quantity 25 g
Molecular Weight (g/mol) 442.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.