Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)-1-(2-thienyl)-1-propanol 98.0+%, TCI America™
SDP

Catalog No. D40091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40091G 1 g
D40095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40091G Supplier TCI America Supplier No. D40091G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13636-02-7
Molecular Formula C9H15NOS
Molecular Weight (g/mol) 185.29
MDL Number MFCD09701963,MFCD07782107
InChI Key XWCNSHMHUZCRLN-UHFFFAOYNA-N
Synonym 2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene
PubChem CID 11586409
IUPAC Name 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol
SMILES CN(C)CCC(O)C1=CC=CS1

Specifications

Melting Point 72°C
Color White
Boiling Point 290°C
Quantity 1 g
Formula Weight 185.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)-1-(2-thienyl)-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.