Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)-1-(2-thienyl)-1-propanone Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D40115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40115G Supplier TCI America Supplier No. D40115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5424-47-5
Molecular Formula C9H14ClNOS
Molecular Weight (g/mol) 219.73
MDL Number MFCD00126349
InChI Key HPVHJPMLORARSR-UHFFFAOYSA-N
Synonym 3-dimethylamino-1-thiophen-2-yl propan-1-one hydrochloride, 3-dimethylamino-1-2-thienyl-1-propanone hydrochloride, 3-dimethylamino-1-2-thienyl-1-propanone hcl, 1-propanone, 3-dimethylamino-1-2-thienyl-, hydrochloride, 2-thienyl 2-dimethylamino ethyl ketone hydrochloride, acmc-209ler, cambridge id 5927860, 2-thienyl-2-dimethylaminoethyl ketone hcl, 2-thienyl 2-dimethylaminoethyl ketone hydrochloride
PubChem CID 2876891
IUPAC Name hydrogen 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-one chloride
SMILES [H+].[Cl-].CN(C)CCC(=O)C1=CC=CS1

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.73
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dimethylamino)-1-(2-thienyl)-1-propanone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.