Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)-1-propanol 98.0+%, TCI America™
SDP

Catalog No. D066125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D066125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D066125ML Supplier TCI America Supplier No. D066125ML
Add to Cart
Add to Cart

Specifications

CAS 3179-63-3
Color Colorless
Boiling Point 164°C
Molecular Formula C5H13NO
Quantity 25 mL
MDL Number MFCD00002945
UN Number 2920
Synonym 3-dimethylamino-1-propanol, 3-dimethylamino propan-1-ol, 1-propanol, 3-dimethylamino, dimethylaminopropanol, n,n-dimethylpropanolamine, 3-dimethylamino propanol, dimethylpropanolamine, 3-dimethlyamino propanol, 3-n,n-dimethylamino propanol
InChI Key PYSGFFTXMUWEOT-UHFFFAOYSA-N
SMILES CN(C)CCCO
IUPAC Name 3-(dimethylamino)propan-1-ol
Molecular Weight (g/mol) 103.17
PubChem CID 76646
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)-1-propanol
Show More Show Less
EINECSNumber (2)-0328
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.