Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Dimethylamino)-1-propylamine

Catalog No. 39380250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-380-250ML 250 mL
39-380-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-380-250ML Supplier Sigma Aldrich Supplier No. 39380250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-(dimethylamino)-1-propylamine (dmapa, n,n-dimethyl-1,3-propanediamine) is a dialkylamino alkyl amine. it has been prepared from dimethylamine and acrylonitrile and confirmed by 1h NMR. it participates in the synthesis of bio-based zwitterionic surfactants from waste cooking oil (wco).3-(dimethylamino)-1-propylamine (dmapa) is suitable reagent used in the following studies:
  • As a cationic moiety to form the ionic liquid, 3-(dimethylamino)-1-propylaminium formate ([dmapa]fa) which is a potential reagent to extract protein from yeast cells
  • As an aminating agent in the palladium-catalyzed amination of aryl amines
  • In the derivatization of pullulan to form cationic pullulan
  • In the partial modification of poly(styrene-alt-maleic anhydride) to form n-substituted maleimide
  • As a test compound in the parallel synthesis of alkylamine lipidoids
  • To cleave the polyamides synthesized by solid phase methods on boc-α-ala-pam resin. it may be used in the following studies:
  • Synthesis of (n,n-dialkylamino)alkyl-2-amino benzamide by reacting with isatoic anhydride
  • As a reagent for the anomeric deacetylation of protected sugars
  • Preparation of bioreducible copolymers used in gene delivery.industrial applications

Specifications

CAS 109-55-7
Density 0.812 g/mL (at 25°C (literature))
Boiling Point 133°C (lit.)
Molecular Formula C5H14N2
Refractive Index n20/D 1.435 (literature); n20/D 1.436
Linear Formula (CH3)2 N(CH2)3 NH2
MDL Number MFCD00008216
Quantity 250 mL
Synonym N,N-Dimethyl-1,3-diaminopropane; N,N-Dimethyl-1,3-propanediamine
Molecular Weight (g/mol) 102.18
Percent Purity ≥98.0% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.