Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Dimethylamino)-1-propylamine

Catalog No. D1450091L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1450091L 1 L
D145009 500 mL
D14500925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1450091L Supplier Sigma Aldrich Supplier No. D1450091L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-(dimethylamino)-1-propylamine is widely used to functionalize various pharmacologically active medicinally important compounds
  • It is also used in the preparation of dimethylaminopropylamine surfactants that exhibit biological activity against gram-positive and negative bacteria and fungi

Specifications

CAS 109-55-7
Density 0.812 g/mL (at 25°C (literature))
Boiling Point 133°C (lit.)
Molecular Formula C5H14N2
Refractive Index n20/D 1.435 (literature)
Linear Formula (CH3)2 N(CH2)3 NH2
Quantity 1 L
MDL Number MFCD00008216
Synonym N,N-Dimethyl-1,3-diaminopropane; N,N-Dimethyl-1,3-propanediamine
Molecular Weight (g/mol) 102.18
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.