Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(dimethylamino)-1-(thiophen-2yl)propan1-ol | 13636-02-7 | MFCD09701963 | 99.8% | 185.29 | 100 MG
SDP

Catalog No. 5000404807
Encompass_Preferred

3-(Dimethylamino)-1-(thiophen-2yl)propan1-ol is classified as a drug intermediate, primarily utilized for the synthesis of various active compounds.

  • Drug intermediate
  • Used for synthesis of various active compounds
  • Purity of 99.8%
  • Appearance: Solid
  • Color: White to off-white
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-04807 Supplier Medchemexpress LLC Supplier No. HY40257100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(Dimethylamino)-1-(thiophen-2yl)propan1-ol is classified as a drug intermediate, primarily utilized for the synthesis of various active compounds.

  • Drug intermediate
  • Used for synthesis of various active compounds
  • Purity of 99.8%
  • Appearance: Solid
  • Color: White to off-white
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.