Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)benzyl alcohol, 95%

Catalog No. AC438160010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438160010 1 g
AC438160050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438160010 Supplier Thermo Scientific Chemicals Supplier No. 438160010
Add to Cart

CAS: 23501-93-1 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23501-93-1
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00017342
InChI Key YTUXZVSDDHCTBZ-UHFFFAOYSA-N
Synonym 3-dimethylamino phenyl methanol, 3-dimethylamino benzyl alcohol, 3-dimethylaminobenzyl alcohol, m-dimethylamino benzyl alcohol, benzenemethanol,3-dimethylamino, 3-dimethylamino phenyl methan-1-ol, benzyl alcohol, m-dimethylamino, benzenemethanol, 3-dimethylamino
PubChem CID 90133
IUPAC Name [3-(dimethylamino)phenyl]methanol
SMILES CN(C)C1=CC=CC(=C1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 109.0°C to 110.0°C (0.1 Torr)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 151.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)benzyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 245-700-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.