Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Dimethylamino)propyl acrylate

Catalog No. 41565025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-565-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-565-025ML Supplier Sigma Aldrich Supplier No. 41565025ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(Dimethylamino)propyl acrylate

Specifications

CAS 18526-07-3
Density 0.928 g/mL (at 25°C)
Boiling Point 167°C
Molecular Formula C8H15NO2
Linear Formula H2C=CHCO2(CH2)3 N(CH3)2
Refractive Index n20/D 1.44 (literature)
MDL Number MFCD00216567
Quantity 25 mL
Synonym 3-(N ,N -Dimethylamino)propyl acrylate; N ,N -Dimethyl-3-aminopropyl acrylate
Molecular Weight (g/mol) 157.21
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.