Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-3-(Dimethylamino)Pyrrolidine

Catalog No. 6567121G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-671-21G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 65-671-21G Supplier Sigma Aldrich Supplier No. 6567121G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(+)-3-(Dimethylamino)Pyrrolidine

Specifications

CAS 871126-22-6
Melting Point 161°C to 178°C
Molecular Formula C7H6BFO3
Refractive Index n20/D 1.4650
Linear Formula C7H6BFO3
MDL Number MFCD07367058
Quantity 10 g
Synonym (3R)-N,N-Dimethyl-3-pyrrolidinamine
Molecular Weight (g/mol) 167.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.