Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dimethylaminophenol, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
Packaging:
Glass bottle
99-07-0
C8H11NO
137.18
MFCD00002264
MESJRHHDBDCQTH-UHFFFAOYSA-N
3-dimethylamino phenol,phenol, 3-dimethylamino,m-dimethylamino phenol,m-dimethylaminophenol,n,n-dimethyl-m-aminophenol,n,n-dimethyl-3-aminophenol,phenol, m-dimethylamino,3-hydroxyphenyl dimethylamine,3-hydroxy-n,n-dimethylaniline,3-n,n-dimethylaminophenol
7421
3-(dimethylamino)phenol
CN(C)C1=CC(=CC=C1)O
99-07-0
C8H11NO
137.18
MFCD00002264
MESJRHHDBDCQTH-UHFFFAOYSA-N
3-dimethylamino phenol,phenol, 3-dimethylamino,m-dimethylamino phenol,m-dimethylaminophenol,n,n-dimethyl-m-aminophenol,n,n-dimethyl-3-aminophenol,phenol, m-dimethylamino,3-hydroxyphenyl dimethylamine,3-hydroxy-n,n-dimethylaniline,3-n,n-dimethylaminophenol
7421
3-(dimethylamino)phenol
CN(C)C1=CC(=CC=C1)O
Melting Point 80°C to 85°C
Flash Point 148°C
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown to Gray or Purple
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)2NC6H4OH
Beilstein 13, 405
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 137.18
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 3-Dimethylaminophenol
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.