Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dimethylaminopropylchloride Hydrochloride 99%, ACROS Organics™

Click to view available options
Quantity:
500g
Packaging:
Glass bottle
5407-04-5
C5H12ClN·HCl
158.07
MFCD00012521
LJQNMDZRCXJETK-UHFFFAOYSA-N
3-dimethylaminopropylchloride hydrochloride,3-chloro-n,n-dimethylpropan-1-amine hydrochloride,3-dimethylaminopropyl chloride hydrochloride,3-dimethylamino propyl chloride hydrochloride,n,n-dimethyl-3-chloropropylamine hydrochloride,3-chloropropyl dimethylamine hydrochloride,3-chloro-n,n-dimethylpropylamine hydrochloride,dimethylaminopropyl chloride hydrochloride,ccris 7054,dimethylaminopropyl chloride, hydrochloride
94308
3-chloro-N,N-dimethylpropan-1-amine;hydrochloride
CN(C)CCCCl.Cl
5407-04-5
C5H12ClN·HCl
158.07
MFCD00012521
LJQNMDZRCXJETK-UHFFFAOYSA-N
3-dimethylaminopropylchloride hydrochloride,3-chloro-n,n-dimethylpropan-1-amine hydrochloride,3-dimethylaminopropyl chloride hydrochloride,3-dimethylamino propyl chloride hydrochloride,n,n-dimethyl-3-chloropropylamine hydrochloride,3-chloropropyl dimethylamine hydrochloride,3-chloro-n,n-dimethylpropylamine hydrochloride,dimethylaminopropyl chloride hydrochloride,ccris 7054,dimethylaminopropyl chloride, hydrochloride
94308
3-chloro-N,N-dimethylpropan-1-amine;hydrochloride
CN(C)CCCCl.Cl
Loss on Drying 1% max. (105°C, 3 hrs)
Melting Point 139.0°C to 145.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500g
Color White to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2N(CH2)3Cl·HCl
Beilstein 04, 148
Solubility Information Solubility in water: soluble 2000g/L in water (20°C). Other solubilities: soluble in ethanol, very soluble in methanol, insoluble in toluene and acetone
Formula Weight 158.07
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 3-Dimethylaminopropylchloride hydrochloride, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 226-467-2

RTECSNumber : UI0209950

TSCA : TSCA