Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Diphenylphosphino)propylamine, 97+%

Catalog No. aah60672md
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60672MD 250 mg
AAH6067203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60672MD Supplier Thermo Scientific Chemicals Supplier No. H60672MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 16605-03-1 | C15H18NP | 243.29 g/mol

3-(Diphenylphosphino)propylamine is used as a paharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Diphenylphosphino)propylamine is used as a paharmaceutical.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16605-03-1
Molecular Formula C15H18NP
Molecular Weight (g/mol) 243.29
MDL Number MFCD00233832
InChI Key DQZWMOWSTWWMPP-UHFFFAOYSA-N
Synonym 3-diphenylphosphino-1-propylamine, 3-diphenylphosphino propan-1-amine, 3-aminopropyl diphenylphosphine, 3-diphenylphosphino propylamine, 3-diphenylphosphanyl-propylamine, 1-propanamine,3-diphenylphosphino, 3-aminopropyl diphenylphosphane, 3-diphenylphosphanyl propan-1-amine, acmc-20aniq, 3-aminopropyldiphenylphosphine
PubChem CID 280677
IUPAC Name 3-diphenylphosphanylpropan-1-amine
SMILES NCCCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Linear Formula (C6H5)2PCH2CH2CH2NH2
Quantity 250 mg
UN Number UN2735
Beilstein 2730850
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 243.28
Percent Purity ≥97%
Chemical Name or Material 3-(Diphenylphosphino)propylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.