Learn More
Ace Glass, Inc. 3 dram poly-seal vial, cs/1152, sp/144
SDP
Catalog No. 50979876
Encompass
3 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention. Pack/144.
Catalog No. 50-979-876 Supplier Ace Glass, Inc. Supplier No. 877930
3 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention. Pack/144.