Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-endo-Aminobicyclo[2.2.1]heptane-2-endo-carboxylic Acid 98%, Thermo Scientific™

Catalog No. AC330242500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC330242500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC330242500 Supplier Thermo Scientific Chemicals Supplier No. 330242500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104308-53-4
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.212
InChI Key KGMHNVXXWOSXSF-VSLZZJKVSA-N
Synonym cis-endo-3-aminobicyclo 2.2.1 heptane-2-carboxylic acid monohydrate
PubChem CID 133655423
IUPAC Name (1R,2R,3S,4S)-3-aminobicyclo[2.2.1]heptane-2-carboxylic acid;hydrate
SMILES C1CC2CC1C(C2N)C(=O)O.O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Quantity 250 mg
Formula Weight 155.19
Percent Purity ≥97.5%
Physical Form Granular Powder
Chemical Name or Material 3-endo-Aminobicyclo[2.2.1]heptane-2-endo-carboxylic acid, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.