Learn More
Cayman Chemical 3-epi Ursolic Acid, Purity: ≥98%, CAS: 989-30-0, 10 mg
SDP
Catalog No. 502912324
Encompass_Preferred
A pentacyclic triterpenoid; inhibits glycogen phosphorylase and cathepsin L but not cathepsin B (IC50s = 19, 6.5, and >250 µM, respectively); inhibits entry of bovine parainfluenza virus 3 into MDBK cells at 20-50 µM; inhibits the proliferation of MCF-7 cells (IC50 = 18.6 µg/ml)
Catalog No. 50-291-2324 Supplier Cayman Chemical Supplier No. 3652810 MG
A pentacyclic triterpenoid; inhibits glycogen phosphorylase and cathepsin L but not cathepsin B (IC50s = 19, 6.5, and >250 µM, respectively); inhibits entry of bovine parainfluenza virus 3 into MDBK cells at 20-50 µM; inhibits the proliferation of MCF-7 cells (IC50 = 18.6 µg/ml)