Learn More
Medchemexpress LLC 3-epi-Vitamin D3 (Epicholecalciferol) | 57651-82-8 | C27H44O | 100 MG
SDP
Catalog No. 5000376787
Encompass_Preferred
3-epi-Vitamin D3 (Epicholecalciferol) is a Vitamin D3 analogue that acts as a Hedgehog pathway inhibitor. It has demonstrated antiproliferative activity in human U87MG cells with an IC50 of 39.2 μM. This compound is intended for research use only.
  • Hedgehog pathway inhibitor
  • Exhibits antiproliferative activity against U87MG cells
  • Significantly upregulates Cyp24A1 in various cell lines
Catalog No. 50-003-76787 Supplier Medchemexpress LLC Supplier No. HY15398C100MG
3-epi-Vitamin D3 (Epicholecalciferol) is a Vitamin D3 analogue that acts as a Hedgehog pathway inhibitor. It has demonstrated antiproliferative activity in human U87MG cells with an IC50 of 39.2 μM. This compound is intended for research use only.
  • Hedgehog pathway inhibitor
  • Exhibits antiproliferative activity against U87MG cells
  • Significantly upregulates Cyp24A1 in various cell lines