Learn More
Medchemexpress LLC 3-epi-Vitamin D3 (Epicholecalciferol) | 57651-82-8 | C27H44O | 25 MG
SDP
Catalog No. 5000374093
Encompass_Preferred
3-epi-Vitamin D3 (Epicholecalciferol) is a Vitamin D3 analogue. It acts as a Hedgehog pathway inhibitor with an IC50 of 39.2 μM in U87MG cells and significantly upregulates Cyp24A1 in U87MG, HT-29, and C3H10T1/2 cells.
  • Acts as a Hedgehog pathway inhibitor
  • Upregulates Cyp24A1 in U87MG, HT-29, and C3H10T1/2 cells
  • Appears as a white to light yellow solid
Catalog No. 50-003-74093 Supplier Medchemexpress LLC Supplier No. 25MGHY15398C
3-epi-Vitamin D3 (Epicholecalciferol) is a Vitamin D3 analogue. It acts as a Hedgehog pathway inhibitor with an IC50 of 39.2 μM in U87MG cells and significantly upregulates Cyp24A1 in U87MG, HT-29, and C3H10T1/2 cells.
  • Acts as a Hedgehog pathway inhibitor
  • Upregulates Cyp24A1 in U87MG, HT-29, and C3H10T1/2 cells
  • Appears as a white to light yellow solid