Learn More
Medchemexpress LLC 3-epi-Vitamin D3 (Epicholecalciferol) | 57651-82-8 | C27H44O | 50 MG
SDP
Catalog No. 5000376508
Encompass_Preferred
3-epi-Vitamin D3 (Epicholecalciferol) is an analogue of Vitamin D3. It functions as a Hedgehog pathway inhibitor with an IC50 of 39.2 μM in U87MG cells. This product is intended for research use only and is not for patient use.
  • Acts as a Hedgehog pathway inhibitor
  • Exhibits antiproliferative activity against U87MG cells
  • Significantly upregulates Cyp24A1 in U87MG, HT-29 and C3H10T1/2 cells
Catalog No. 50-003-76508 Supplier Medchemexpress LLC Supplier No. HY15398C50MG
3-epi-Vitamin D3 (Epicholecalciferol) is an analogue of Vitamin D3. It functions as a Hedgehog pathway inhibitor with an IC50 of 39.2 μM in U87MG cells. This product is intended for research use only and is not for patient use.
  • Acts as a Hedgehog pathway inhibitor
  • Exhibits antiproliferative activity against U87MG cells
  • Significantly upregulates Cyp24A1 in U87MG, HT-29 and C3H10T1/2 cells