Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-epi-vitamin D3 (epicholecalciferol) | 57651-82-8 | MFCD28963703 | 94.1% | 384.64 g·mol⁻1 | C27H44O | 10 MG
SDP

Catalog No. 5000285807
Encompass_Preferred

3-epi-Vitamin D3 (Epicholecalciferol) is a vitamin D3 analogue used in research to modulate the Hedgehog signaling pathway. It exhibits antiproliferative activity in human U87MG cells (IC50 = 39.2 μM, MTS assay, 72 h) and is provided as a purified small-molecule powder for in vitro studies.

  • Molecular formula C27H44O.
  • Molecular weight 384.64 g·mol⁻1.
  • CAS number 57651-82-8.
  • Purity 94.1%.
  • Bioactivity Hedgehog pathway inhibition; IC50 = 39.2 μM in U87MG cells (MTS assay, 72 h).
  • Supplied in milligram pack sizes suitable for cell-based assays.

Catalog No. 50-002-85807 Supplier Medchemexpress LLC Supplier No. HY15398C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-epi-Vitamin D3 (Epicholecalciferol) is a vitamin D3 analogue used in research to modulate the Hedgehog signaling pathway. It exhibits antiproliferative activity in human U87MG cells (IC50 = 39.2 μM, MTS assay, 72 h) and is provided as a purified small-molecule powder for in vitro studies.

  • Molecular formula C27H44O.
  • Molecular weight 384.64 g·mol⁻1.
  • CAS number 57651-82-8.
  • Purity 94.1%.
  • Bioactivity Hedgehog pathway inhibition; IC50 = 39.2 μM in U87MG cells (MTS assay, 72 h).
  • Supplied in milligram pack sizes suitable for cell-based assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.