Learn More
Cayman Chemical 3-Epideoxycholic Acid, Purity: ≥98%, CAS: 570-63-8, 10 mg
SDP
Catalog No. 502913127
Encompass_Preferred
A secondary bile acid produced by gut microbiota; increases the growth of Caco-2 cells at 10 and 20 µM; increases Foxp3+CD4+ Treg levels in a co-culture of naïve T cells and dendritic cells
Catalog No. 50-291-3127 Supplier Cayman Chemical Supplier No. 3498410 MG
A secondary bile acid produced by gut microbiota; increases the growth of Caco-2 cells at 10 and 20 µM; increases Foxp3+CD4+ Treg levels in a co-culture of naïve T cells and dendritic cells