Learn More
Medchemexpress LLC 3-Epideoxycholic acid | 570-63-8 | 99.9% | C24H40O4 | 1 MG
SDP
Catalog No. 5000357770
Encompass_Preferred
3-Epideoxycholic acid is a microbial metabolite that targets FXR of dendritic cells. It reduces immunostimulatory properties, promotes the generation of Treg cells, and exhibits anti-inflammatory activity. Additionally, it promotes the growth of Bacteroides bacteria.
  • Targets FXR of dendritic cells
  • Reduces immunostimulatory properties
  • Promotes generation of Treg cells
  • Exhibits anti-inflammatory activity
  • Promotes growth of Bacteroides bacteria
  • Functions as an endogenous metabolite
Catalog No. 50-003-57770 Supplier Medchemexpress LLC Supplier No. HY488141MG
Add to Cart
Add to Cart
3-Epideoxycholic acid is a microbial metabolite that targets FXR of dendritic cells. It reduces immunostimulatory properties, promotes the generation of Treg cells, and exhibits anti-inflammatory activity. Additionally, it promotes the growth of Bacteroides bacteria.
  • Targets FXR of dendritic cells
  • Reduces immunostimulatory properties
  • Promotes generation of Treg cells
  • Exhibits anti-inflammatory activity
  • Promotes growth of Bacteroides bacteria
  • Functions as an endogenous metabolite