Learn More
Medchemexpress LLC 3-Epideoxycholic acid | 570-63-8 | 99.9% | C24H40O4 | 5 MG
SDP
Catalog No. 5000359251
Encompass_Preferred
3-Epideoxycholic acid is a microbial metabolite of Deoxycholic acid. It targets FXR of dendritic cells, which reduces their immunostimulatory properties and promotes the generation of Treg cells, showing anti-inflammatory activity.
  • Targets FXR of dendritic cells
  • Reduces immunostimulatory properties of dendritic cells
  • Promotes generation of Treg cells
  • Exhibits anti-inflammatory activity
  • Promotes the growth of Bacteroides bacteria
Catalog No. 50-003-59251 Supplier Medchemexpress LLC Supplier No. HY488145MG
Add to Cart
Add to Cart
3-Epideoxycholic acid is a microbial metabolite of Deoxycholic acid. It targets FXR of dendritic cells, which reduces their immunostimulatory properties and promotes the generation of Treg cells, showing anti-inflammatory activity.
  • Targets FXR of dendritic cells
  • Reduces immunostimulatory properties of dendritic cells
  • Promotes generation of Treg cells
  • Exhibits anti-inflammatory activity
  • Promotes the growth of Bacteroides bacteria