Learn More
Medchemexpress LLC HY-N4289 5mg Medchemexpress, 3-Epiursolic Acid CAS:989-30-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN42895MG
Encompass_Preferred
Medchemexpress, HY-N4289 5mg 3-Epiursolic Acid CAS:989-30-0 3-Epiursolic Acid is a triterpenoid isolated from Myrtaceae, acts as a competitive inhibitor of cathepsin L (ICIC50, 6.5 μM; Ki, 19.5 μM), with no obvious effect on cathepsin B. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0438