Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Epiursolic Acid | 989-30-0 | 1 MG
SDP

Catalog No. 5000418671
Encompass_Preferred

3-Epiursolic Acid is a triterpenoid isolated from Eriobotrya japonica. It acts as a competitive inhibitor of cathepsin L with an IC50 of 6.5 μM and a Ki of 19.5 μM. It has no significant effect on cathepsin B. This product is for research use only.

  • Purity: 99.77%
  • Acts as a competitive inhibitor of cathepsin L (IC50: 6.5 μM)
  • Appearance: White to off-white solid
  • Molecular formula: C30H48O3
  • Molecular weight: 456.70
  • Structure classification: Terpenoids, triterpenes
  • Initial source: Plants (Rosaceae, Eriobotrya japonica (Thunb.) Lindl.)

Catalog No. 50-004-18671 Supplier Medchemexpress LLC Supplier No. HYN42891MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Epiursolic Acid is a triterpenoid isolated from Eriobotrya japonica. It acts as a competitive inhibitor of cathepsin L with an IC50 of 6.5 μM and a Ki of 19.5 μM. It has no significant effect on cathepsin B. This product is for research use only.

  • Purity: 99.77%
  • Acts as a competitive inhibitor of cathepsin L (IC50: 6.5 μM)
  • Appearance: White to off-white solid
  • Molecular formula: C30H48O3
  • Molecular weight: 456.70
  • Structure classification: Terpenoids, triterpenes
  • Initial source: Plants (Rosaceae, Eriobotrya japonica (Thunb.) Lindl.)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.