Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Ethoxy-3-Oxopropylzinc Bromide Solution

Catalog No. 49852150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-852-150ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 49-852-150ML Supplier Sigma Aldrich Supplier No. 49852150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Ethoxy-3-oxopropylzinc bromide is a reagent used to insert ethyl propanoate group to the substrate. It can be used in:Synthesis of +a-keto esters from aryl chlorides.Synthesis of sulfones from 1,4-diazabicyclo[2.2.2]octane bis(sulfur dioxide) (DABSO) and alkyl halides.Negishi cross-couplings with aryl vinyl phosphates to synthesize 1,1-disubstituted alkenes.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 312693-01-9
Density 0.98 g/mL at 25°C
Molecular Formula C8H15BrO2Zn
Linear Formula C2H5O2C(CH2)5ZnBr
MDL Number MFCD00671979
Quantity 50 mL
Molecular Weight (g/mol) 288.5 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.