Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxyacrylic acid, 95%, Thermo Scientific™

Catalog No. RH00924DE
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RH00924DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RH00924DE Supplier Thermo Scientific Chemicals Supplier No. RH00924DE
Add to Cart

Chemical Identifiers

CAS 6192-01-4
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
InChI Key SYMAGJYJMLUEQE-ONEGZZNKSA-N
Synonym 3-ethoxyacrylic acid, e-3-ethoxyacrylic acid, 3-ethoxyprop-2-enoic acid, 3-ethyoxyacrylic acid, 3-ethoxy-2-propenoic acid, 2e-3-ethoxyprop-2-enoic acid, 2e-3-ethoxyacrylic acid, 3-ethoxyacrylicacid, pubchem20958, e-3-ethoxy-acrylic acid
PubChem CID 5709609

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 116.12
Percent Purity 95%
Chemical Name or Material 3-Ethoxyacrylic acid
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.