Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxybenzenethiol 98.0+%, TCI America™
SDP

Catalog No. E07011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07011G Supplier TCI America Supplier No. E07011G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 86704-82-7
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD03094660
InChI Key RTJMJGJLVSKSEB-UHFFFAOYSA-N
Synonym 3-Ethoxythiophenol
PubChem CID 2736350
IUPAC Name 3-ethoxybenzene-1-thiol
SMILES CCOC1=CC(S)=CC=C1

Specifications

Color Yellow
Boiling Point 239°C
Quantity 1 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethoxybenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.