Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Ethoxycarbonyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AC445020010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445020010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445020010 Supplier Thermo Scientific Chemicals Supplier No. 445020010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 269410-00-6
Molecular Formula C15H21BO4
Molecular Weight (g/mol) 276.14
MDL Number MFCD03093896
InChI Key RQKGHIGGSFNVGW-UHFFFAOYSA-N
Synonym 3-Carboethoxyphenylboronic acid pinacol ester
IUPAC Name ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES CCOC(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 41°C to 43°C
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 276.14
Percent Purity 97%
Chemical Name or Material 3-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.