Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E07241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07241G 1 g
E0724-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07241G Supplier TCI America Supplier No. E07241G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90555-66-1
Molecular Formula C8H11BO3
MDL Number MFCD00274219
InChI Key CHCWUTJYLUBETR-UHFFFAOYSA-N
PubChem CID 2773920
IUPAC Name (3-ethoxyphenyl)boronic acid

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 1 g
Synonym 3-ethoxyphenyl boronic acid, 3-ethoxybenzeneboronic acid, 3-ethoxyphenylboronicacid, 3-ethoxyphenyl boranediol, m-ethoxyphenylboronic acid, boronic acid, 3-ethoxyphenyl, pubchem1835, acmc-209r6q, 3-ethoxy-phenylboronic acid
SMILES B(C1=CC(=CC=C1)OCC)(O)O
Molecular Weight (g/mol) 165.983
Formula Weight 165.98
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.