Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E07245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0724-5G 5 g
E07241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0724-5G Supplier TCI America Supplier No. E07245G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90555-66-1
Molecular Formula C8H11BO3
MDL Number MFCD00274219
InChI Key CHCWUTJYLUBETR-UHFFFAOYSA-N
PubChem CID 2773920
IUPAC Name (3-ethoxyphenyl)boronic acid

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 5 g
Synonym 3-ethoxyphenyl boronic acid, 3-ethoxybenzeneboronic acid, 3-ethoxyphenylboronicacid, 3-ethoxyphenyl boranediol, m-ethoxyphenylboronic acid
SMILES CCOC1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 165.98
Formula Weight 165.98
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.