Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethyl-2-pentene 98.0+%, TCI America™
SDP

Catalog No. E01585ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E01585ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E01585ML Supplier TCI America Supplier No. E01585ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 816-79-5
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00027079
InChI Key XMYFZAWUNVHVGI-UHFFFAOYSA-N
Synonym 1,1-Diethyl-1-propene
PubChem CID 13159
IUPAC Name 3-ethylpent-2-ene
SMILES CCC(=CC)CC

Specifications

Color Colorless
Boiling Point 96°C
Quantity 5 mL
UN Number 3295
Formula Weight 98.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethyl-2-pentene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.