Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethyl-3-methylglutaric Acid 98.0+%, TCI America™
SDP

Catalog No. E02855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02855G Supplier TCI America Supplier No. E02855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5345-01-7
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00020494
InChI Key XAWFHZMTJUGGEE-UHFFFAOYSA-N
PubChem CID 79290
IUPAC Name 3-ethyl-3-methylpentanedioic acid
SMILES CCC(C)(CC(=O)O)CC(=O)O

Specifications

Melting Point 83°C
Color White
Quantity 5 g
Formula Weight 174.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl-3-methylglutaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.