Learn More
Medchemexpress LLC 3-ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1-(prop-2-yn-1-yl)-1,4-dihydropyridine-3,5-dicarbo | 544478-19-5 | MFCD06798381 | 99.5% | 398.41 g/mol | C21H22N2O6 | 100 MG
SDP
Catalog No. 5000244906
Encompass_Preferred
MRS1845 is a small-molecule research reagent that inhibits store-operated calcium (SOC) channels and ORAI1, used to study SOCE-mediated calcium entry in vitro. It is supplied as a solid powder with reported molecular formula C21H22N2O6 and a molecular weight of 398.41 g/mol.
  • Selective SOC/ORAI1 inhibition with reported IC50 ≈ 1.7 μM.
  • High reported purity suitable for research applications.
  • Supplied as a solid powder for solution preparation and cell assays.
  • Stable under recommended storage conditions for extended shelf life.
Catalog No. 50-002-44906 Supplier Medchemexpress LLC Supplier No. HY103310100MG
MRS1845 is a small-molecule research reagent that inhibits store-operated calcium (SOC) channels and ORAI1, used to study SOCE-mediated calcium entry in vitro. It is supplied as a solid powder with reported molecular formula C21H22N2O6 and a molecular weight of 398.41 g/mol.
  • Selective SOC/ORAI1 inhibition with reported IC50 ≈ 1.7 μM.
  • High reported purity suitable for research applications.
  • Supplied as a solid powder for solution preparation and cell assays.
  • Stable under recommended storage conditions for extended shelf life.