Learn More
Medchemexpress LLC 3-ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1-(prop-2-yn-1-yl)-1,4-dihydropyridine-3,5-dicarboxylat | 544478-19-5 | MFCD06798381 | 99.5% | 398.41 g/mol | C21H22N2O6 | 25 MG
SDP
Catalog No. 5000244841
Encompass_Preferred
MRS1845 is a small-molecule inhibitor of store-operated calcium entry (SOCE) that targets ORAI1 channels. It is used in cellular research to probe SOCE/ORAI1-mediated calcium signaling and calcium entry mechanisms. Reported activity includes an IC50 of 1.7 μM in cellular assays.
  • Inhibits store-operated calcium entry (SOCE) and targets ORAI1.
  • Reported IC50 of 1.7 μM in HL-60 cellular assays.
  • Molecular formula C21H22N2O6; molecular weight 398.41 g/mol.
  • Reported purity approximately 99.46% (manufacturer listing).
  • Available as solid packs (including 25 mg) and as a 10 mM solution in DMSO for ready reconstitution.
Catalog No. 50-002-44841 Supplier Medchemexpress LLC Supplier No. HY10331025MG
MRS1845 is a small-molecule inhibitor of store-operated calcium entry (SOCE) that targets ORAI1 channels. It is used in cellular research to probe SOCE/ORAI1-mediated calcium signaling and calcium entry mechanisms. Reported activity includes an IC50 of 1.7 μM in cellular assays.
  • Inhibits store-operated calcium entry (SOCE) and targets ORAI1.
  • Reported IC50 of 1.7 μM in HL-60 cellular assays.
  • Molecular formula C21H22N2O6; molecular weight 398.41 g/mol.
  • Reported purity approximately 99.46% (manufacturer listing).
  • Available as solid packs (including 25 mg) and as a 10 mM solution in DMSO for ready reconstitution.