Learn More
Sigma Aldrich Fine Chemicals Biosciences 3-(Ethylenediamino)propyl-functionalized silica gel 200-400 mesh, extent of labeling: 1.4 mmol/g loading | 1173023-01-2 | MFCD03095961 | 5G
Catalog No. 501752065
Encompass
3-(Ethylenediamino)propyl-functionalized silica gel 200-400 mesh, extent of labeling: 1.4 mmol/g loading | 1173023-01-2 | MFCD03095961 | 5G
Catalog No. 50-175-2065 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5379185G
3-(Ethylenediamino)propyl-functionalized silica gel 200-400 mesh, extent of labeling: 1.4 mmol/g loading | 1173023-01-2 | MFCD03095961 | 5G