Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Ethylphenylboronic acid

Catalog No. SYX000371G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX000371G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX000371G Supplier Sigma Aldrich Supplier No. SYX000371G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,3,4,5,6-pentafluorobenzonitrile was used in the synthesis of novel fluorinated poly(ether nitrile)s
  • It was also used in the synthesis of pentafluorophenyl analogs of noradrenaline, adrenaline and n-methyladrenaline

Specifications

CAS 90555-65-0
Molecular Formula C8H11BO2
Linear Formula C8H11BO2
MDL Number MFCD06657721
Quantity 10 g
Molecular Weight (g/mol) 149.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.