Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethylpyridine, 98%

Catalog No. AAB2031806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2031806 5 g
AAB2031814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2031806 Supplier Thermo Scientific Chemicals Supplier No. B2031806

CAS: 536-78-7 | C7H9N | 107.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-78-7
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006413
InChI Key MFEIKQPHQINPRI-UHFFFAOYSA-N
Synonym pyridine, 3-ethyl, 3-ethyl pyridine, beta-lutidine, beta-ethylpyridine, 3-ethyl-pyridine, 5-ethylpyridine, unii-a25i3ez88v, lutidine, beta, 3ethylpyridine, fema no. 3394
PubChem CID 10823
IUPAC Name 3-ethylpyridine
SMILES CCC1=CN=CC=C1

Specifications

Melting Point -77°C
Boiling Point 165°C to 166°C
Flash Point 49°C (120°F)
Odor Unpleasant
Quantity 5 g
UN Number UN1993
Beilstein 106479
Merck Index 14,3848
Formula Weight 107.16
Percent Purity 98%
Chemical Name or Material 3-Ethylpyridine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.