Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethylthio-1,2-propanediol 97%, Thermo Scientific™

Catalog No. AC206780250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC206780250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC206780250 Supplier Thermo Scientific Chemicals Supplier No. 206780250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 60763-78-2
CAS Min % 96.0
CAS Max % 100.0
Density 1.09
Infrared Spectrum Authentic
Molecular Formula C5H12O2S
Refractive Index 1.5055 to 1.5075 (20°C, 589nm)
Linear Formula C2H5SCH2CH(OH)CH2OH
Quantity 25g
MDL Number MFCD00004725
Synonym 3-ethylthio-1,2-propanediol, 3-ethylsulfanyl propane-1,2-diol, 3-ethylthiopropane-1,2-diol, 3-ethyl thio-1,2-propanediol, gilaqkilvghfpx-uhfffaoysa, 1,2-propanediol,3-ethylthio, 3-ethylsulfanyl-propane-1,2-diol, 1,2-propanediol, 3-ethylthio, 3-ethylsulfanyl-1,2-propanediol #
InChI Key GILAQKILVGHFPX-UHFFFAOYSA-N
SMILES CCSCC(CO)O
IUPAC Name 3-ethylsulfanylpropane-1,2-diol
Molecular Weight (g/mol) 136.209
PubChem CID 108509
Formula Weight 136.21
Percent Purity ≥96.0%
Chemical Name or Material 3-Ethylthio-1, 2-propanediol, 97%
Show More Show Less
Hazard Category STENCH
EINECSNumber 262-419-7
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.