Learn More
Medchemexpress LLC 3-Ethynylaniline | 54060-30-9 | 117.15 | 1 ML
SDP
Catalog No. 5000460431
Encompass_Preferred
3-Ethynylaniline is a biochemical reagent used as a biological material or organic compound in life science research. It functions as a click chemistry reagent due to its Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. Click chemistry is a method for selectively, modularly, and efficiently synthesizing new compounds through heteroatom linking.
  • Biochemical reagent for life science research
  • Functions as a click chemistry reagent
  • Contains an alkyne group for reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-004-60431 Supplier Medchemexpress LLC Supplier No. HYW00742310MM/1ML
3-Ethynylaniline is a biochemical reagent used as a biological material or organic compound in life science research. It functions as a click chemistry reagent due to its Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. Click chemistry is a method for selectively, modularly, and efficiently synthesizing new compounds through heteroatom linking.
  • Biochemical reagent for life science research
  • Functions as a click chemistry reagent
  • Contains an alkyne group for reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)