Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ferrocenoylpropionic Acid 95.0+%, TCI America™
SDP

Catalog No. F032025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F032025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F032025G Supplier TCI America Supplier No. F032025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1291-72-1
Molecular Formula C14H14FeO3
MDL Number MFCD00027357
Synonym 4-Ferrocenyl-4-oxobutyric Acid

Specifications

Quantity 25 g
Formula Weight 286.11
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ferrocenoylpropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.