Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ferrocenylpropionic anhydride, ≥98.0% (C), MilliporeSigma™ Supelco™

Catalog No. 111012939 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2939 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2939 Supplier MilliporeSigma Supelco Supplier No. 76737100MG
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD18071429
Synonym 1,1 ′′-[Oxybis(3-oxo-3,1-propanediyl)]bis-ferrocene

Specifications

CAS 132098-76-1
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C26H26 Fe2O3
Quantity 100 mg
Formula Weight 498.17
Percent Purity ≥98.0% (C)
Grade For HPLC derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.