Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Feruloyl-4-caffeoylquinic acid | 96990-65-7 | 530.48 | 1 MG
SDP

Catalog No. 5000417156
Encompass_Preferred

3-Feruloyl-4-caffeoylquinic acid is a caffeoyl, feruloyl quinic acid derivative isolated from roasted *C. arabica*. It is intended for research use only.

  • Purity: 97.38%.
  • Appearance: Solid.
  • Color: White to off-white.
  • Structure classification: Phenols, polyphenols.
  • Initial source: Plants, Rubiaceae, *Coffea canephora Pierre ex Froehn.*
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Room temperature, protect from light; in solvent: -80°C for 1 year, -20°C for 6 months.

Catalog No. 50-004-17156 Supplier Medchemexpress LLC Supplier No. HYN100441MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Feruloyl-4-caffeoylquinic acid is a caffeoyl, feruloyl quinic acid derivative isolated from roasted *C. arabica*. It is intended for research use only.

  • Purity: 97.38%.
  • Appearance: Solid.
  • Color: White to off-white.
  • Structure classification: Phenols, polyphenols.
  • Initial source: Plants, Rubiaceae, *Coffea canephora Pierre ex Froehn.*
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Room temperature, protect from light; in solvent: -80°C for 1 year, -20°C for 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.