Learn More
Aobchem 3-Fluoro-2 4-dimethoxybenzoic acid | ≥95% | CAS 1018451-10-9 | C9H9FO4 | 500mg
Catalog No. 5000902544
Encompass_Preferred
3-Fluoro-2 4-dimethoxybenzoic acid | ≥95% | CAS 1018451-10-9 | C9H9FO4 | 500mg
Catalog No. 50-009-02544 Supplier Aobchem Supplier No. A22757500MG
3-Fluoro-2 4-dimethoxybenzoic acid | ≥95% | CAS 1018451-10-9 | C9H9FO4 | 500mg